Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: NT2D1, IMR32.
Predicted reactivity: Mouse (98%), Rat (97%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Guanine nucleotide-binding proteins (G proteins) are modulators or transducers in various transmembrane signaling systems. G proteins are composed of 3 subunits; alpha, beta and gamma. The alpha chain contains the guanine nucleotide binding site.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G alpha-12; G-protein subunit alpha-12; guanine nucleotide binding protein (G protein) alpha 12; Guanine nucleotide-binding protein subunit alpha-12; WUGSC:H_GS165O14.2
Gene Aliases: gep; GNA12; NNX3; RMP
UniProt ID: (Human) Q03113
Entrez Gene ID: (Human) 2768
Molecular Function: heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support