Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 12
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: PDDSGTFYD QAVVSNDMEE HLEEPVAEPE PDPEPEPEQE PVSEIQEEKP EPVLEETAPE DAQKSSSPAP ADIAQTVQED LRTFSWASVT SKNLPPSGAV PVTGIPPHVV KVPASQPRPE SKPESQIPPQ RPQRDQRVRE QRINIPPQRG PRPIREAGEQ GDIEPRRMVR HPDSHQLFIG NLPHEVDKSE LKDFFQSYGN VVELRINSGG KLPNFGFVVF DDSEPVQKVL SNRPIMFRGE VRLNVEEKKT RAAREGDRRD NRLRGPGGPR GGLGGGMRGP PRGGMVQKPG FGVGRGLAPR Q (167-466 aa encoded by BC006997)
This gene encodes one of the DNA-unwinding enzymes which prefers partially unwound 3'-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion. This enzyme is a member of the heterogeneous nuclear RNA-binding proteins and is also an element of the Ras signal transduction pathway. It binds specifically to the Ras-GTPase-activating protein by associating with its SH3 domain. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent DNA helicase VIII; DNA helicase VIII; G3BP; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; HDH-VIII; MGC111040; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RasGAP-associated endoribonuclease G3BP; Rasputin homolog; unnamed protein product
Gene Aliases: B430204O07; G3BP; HDH-VIII; mKIAA4115; RGD1310666
UniProt ID: (Human) Q13283, (Mouse) P97855
Entrez Gene ID: (Human) 10146, (Pig) 100513766, (Mouse) 27041, (Rat) 171092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support