Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GAT-1 Antibodies

          Alomone Labs, LTD

          GABA Transporter 1 (GAT-1) (extracellular) Polyclonal Antibody

          View all (21) GAT-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GABA Transporter 1 (GAT-1) (extracellular) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          GABA Transporter 1 (GAT-1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          GABA Transporter 1 (GAT-1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GABA Transporter 1 (GAT-1) (extracellular) Antibody (AGT-001-GP-200UL) in IHC (F)

          Expression of GABA Transporter 1 in mouse cerebellum. Immunohistochemical staining of perfusion-fixed frozen mouse brain sections with Guinea Pig Anti-GABA Transporter 1 (GAT-1) (extracellular) Antibody (#AGT-001-GP) (1:300), followed by goat anti-guinea pig-AlexaFluor-594. A. GAT-1 immunoreactivity (red) appears in pinceau formations (arrows) around Purkinje cells (P). B. Pre-incubation of the antibody wit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GABA Transporter 1 (GAT-1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GABA Transporter 1 (GAT-1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GABA Transporter 1 (GAT-1) (extracellular) Antibody in Western Blot (WB)
          GABA Transporter 1 (GAT-1) (extracellular) Polyclonal Antibody

          Product Details

          AGT-001-GP-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:300
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Guinea pig / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide KPKESARQDEGKED, corresponding to amino acid residues 205-218 of rat GAP43. C-terminus.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Amount

          160 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Immunogen sequence: KPKESARQDEGKED

          Reconstitution: 25 uL, 50 uL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GABA T-1; GABA T1; GABA transporter 1; GABA transporter protein; GABAT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1; unnamed protein product

          View more View less

          Gene Aliases: A730043E01; GABATHG; GABATR; Gabt; GABT1; GAT-1; GAT1; hGAT-1; MAE; SLC6A1; XT-1; Xtrp1

          View more View less

          UniProt ID: (Human) P30531, (Rat) P23978, (Mouse) P31648

          View more View less

          Entrez Gene ID: (Human) 6529, (Rat) 79212, (Mouse) 232333

          View more View less

          Function(s)
          sodium:amino acid symporter activity gamma-aminobutyric acid:sodium symporter activity protein binding gamma-aminobutyric acid transmembrane transporter activity symporter activity sodium:chloride symporter activity transmembrane transporter activity metal ion binding neurotransmitter:sodium symporter activity primary active transporter
          Process(es)
          amino acid transmembrane transport neurotransmitter transport amino acid transport chemical synaptic transmission memory associative learning sodium ion transmembrane transport synapse organization gamma-aminobutyric acid reuptake gamma-aminobutyric acid import inorganic anion import across plasma membrane sodium ion import across plasma membrane neurotransmitter reuptake transport across blood-brain barrier chloride transmembrane transport learning response to toxic substance response to sucrose response to organic substance response to organonitrogen compound response to lead ion positive regulation of gamma-aminobutyric acid secretion response to organic cyclic compound response to purine-containing compound negative regulation of synaptic transmission, GABAergic response to estradiol response to cocaine protein homooligomerization response to calcium ion transmembrane transport transport mitophagy in response to mitochondrial depolarization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-x2g2k:80/100.66.77.199:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline