Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
1 µg/mL of MA5-45487 was sufficient for detection of alpha1 GABA receptor in 10 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 55kDa. No cross-reactivity against GABA-A-R-Alpha 2 or -Alpha3.
Gamma -aminobutyric acid (GABA) is the primary inhibitory neurotransmitter in the central nervous system, causing a hyperpolarization of the membrane through the opening of a Cl- channel associated with the GABAA Receptor (GABAA-R) subtype. GABAA-Rs are important therapeutic targets for a range of sedative, anxiolytic, and hypnotic agents and are implicated in several diseases including epilepsy, anxiety, depression, and substance abuse. The GABAA-R is a multimeric subunit complex. To date aix alphas, four betas and four gammas, plus alternative splicing variants of some of these subunits, have been identified. Injection in oocytes or mammalian cell lines of cRNA coding for alpha- and beta-subunits results in the expression of functional GABAA-R sensitive to GABA. However, coexpression of a gamma-subunit is required for benzodiazepine modulation. The various effects of the benzodiazepines in brain may also be mediated via different alpha-subunits of the receptor. Lastly, phosphorylation of beta-subunits of the receptor has been shown to modulate GABAA-R function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit alpha-1; GABA(A) receptor, alpha 1; GABAA alpha 1; GABAAR alpha1; GABRA 1; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 1; gamma-aminobutyric acid (GABA) A receptor, alpha 1; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 1; gamma-aminobutyric acid A receptor, alpha 1; Gamma-aminobutyric acid receptor subunit alpha-1
Gene Aliases: ECA4; EIEE19; EJM; EJM5; Gabra-1; GABRA1
UniProt ID: (Human) P14867, (Rat) P62813, (Mouse) P62812
Entrez Gene ID: (Human) 2554, (Rat) 29705, (Mouse) 14394
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support