Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (PFA fixed) (IHC (PFA)) |
1:100 | - |
Immunocytochemistry (ICC/IF) |
Assay-Dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Rat |
Host/Isotype |
Mouse / IgG2A |
Class |
Monoclonal |
Type |
Antibody |
Clone |
S413-67 |
Immunogen |
Fusion protein amino acids 27-253 (Extracellular N-terminus) of human GABA A Receptor Epsilon |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS, pH 7.4, with 50% glycerol |
Contains |
0.1% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2735197 |
The product of this gene belongs to the ligand-gated ionic channel (TC 1.A.9) family. It encodes the gamma-aminobutyric acid (GABA) A receptor which is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes an epsilon subunit. It is mapped to chromosome Xq28 in a cluster comprised of genes encoding alpha 3, beta 4 and theta subunits of the same receptor. Alternatively spliced transcript variants have been identified, but only one is thought to encode a protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) mu Receptor; GABA(A) receptor subunit epsilon; GABA(A) receptor, epsilon; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, epsilon; gamma-aminobutyric acid (GABA-A) receptor, subunit epsilon; gamma-aminobutyric acid A receptor, epsilon; Gamma-aminobutyric acid receptor subunit epsilon
Gene Aliases: GABRE
UniProt ID: (Human) P78334
Entrez Gene ID: (Human) 2564, (Rat) 65191
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support