Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SDDDYEDYAS NKTWVLTPKV PEGDVTVILN NLLEGYDNKL RPDIGVKPTL IHTDMYVNSI GPVNAINMEY TIDIFFAQTW YDRRLKFNST IKVLRLNSNM VGKIWIPDTF FRNSKKADAH WITTPNRMLR IWNDGRVLYT LRLTIDAECQ LQLHNFPMDE HSCPLEFSSY GYPREEIVYQ WKRSSVEVGD TRSWRLYQFS FVGLRNTTEV VKTTSGDYVV MSVYFDLSRR MGY
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit gamma-2; GABAAR subunit gamma-2; GABRA2; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor, gamma 2; Gamma-aminobutyric acid receptor subunit gamma-2; gamma-aminobutyric acid type A receptor gamma2 subunit
Gene Aliases: GABAA-R; Gabrg-2; Gabrg2; gamma2
UniProt ID: (Mouse) P22723, (Rat) P18508
Entrez Gene ID: (Rabbit) 100357302, (Mouse) 14406, (Rat) 29709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support