Sequence of this protein is as follows: PLSEETWHTH QFNFIKNHAF RLLKPGGVLT YCNLTSWGEL MKSKYSDITI MFEETQVPAL LEAGFRRENI RTEVMALVPP ADCRYYAFPQ MITPLVTK
GAMT is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in its gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals.The protein encoded by this gene is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in this gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals. Two transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 20; Guanidinoacetate N-methyltransferase
Gene Aliases: CCDS2; GAMT; HEL-S-20; PIG2; TP53I2
UniProt ID: (Human) Q14353
Entrez Gene ID: (Human) 2593
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support