Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • GATA2 Antibodies

          Invitrogen

          GATA2 Recombinant Rabbit Monoclonal Antibody (PSH04-53)

          Advanced Verification
          View all (22) GATA2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite GATA2 Recombinant Rabbit Monoclonal Antibody (PSH04-53)

          • Antibody Testing Data (6)
          • Advanced Verification (2)
          GATA2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GATA2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GATA2 Antibody (MA562900) in ICC/IF

          Immunofluorescence analysis of GATA2 using SH-SY5Y (positive) and Jurkat (negative). The cells were fixed in 4% paraformaldehyde for 20 minutes at room temperature, permeabilized with 0.1% Triton X-100 in PBS for 5 minutes at room temperature, then blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. The cells were co-labeled with GATA2 Recombinant Rabbit Monoclonal Antibody (PSH04-53) (Product # MA562900) (green) and beta tubulin (red) at 1:100 dilution in 1% BSA in PBST overnight at 4 degrees Celsius and then wi... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GATA2 Antibody in Immunocytochemistry (ICC/IF)
          GATA2 Antibody in Western Blot (WB)
          GATA2 Antibody in Western Blot (WB)
          GATA2 Antibody in Western Blot (WB)
          GATA2 Antibody in Western Blot (WB)
          GATA2 Antibody in Western Blot (WB)
          GATA2 Antibody
          GATA2 Antibody
          GATA2 Recombinant Rabbit Monoclonal Antibody (PSH04-53)

          Product Details

          MA562900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH04-53

          Immunogen

          Synthetic peptide within human GATA2 aa 1-50.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive Control: SH-SY5Y cell lysate, LNCaP cell lysate, PC-12 cell lysate, SH-SY5Y.

          Tissue Specificity: Low tissue specificity.

          Subcellular Location: SH-SY5Y cell lysate, LNCaP cell lysate, PC-12 cell lysate, SH-SY5Y.

          Sequence Similarities: 100% Mouse/Rat.

          Predicted band size: 51 kDa.

          Target Information

          GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Endothelial transcription factor GATA-2; FLJ45948; GATA-binding protein 2; GATA-bindning protein 2; gata2 {ECO:0000312|EMBL:AAK51128.1}; MGC2306

          View more View less

          Gene Aliases: DCML; GATA2; IMD21; MONOMAC; NFE1B

          View more View less

          UniProt ID: (Human) P23769, (Rat) Q924Y4

          View more View less

          Entrez Gene ID: (Human) 2624, (Rat) 25159

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription factor binding enhancer sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding transcription factor binding zinc ion binding C2H2 zinc finger domain binding DNA binding sequence-specific DNA binding DNA-binding transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter urogenital system development cell fate determination neuron migration embryonic placenta development transcription, DNA-templated transcription from RNA polymerase II promoter phagocytosis positive regulation of cytosolic calcium ion concentration heart development blood coagulation organ morphogenesis tissue development regulation of primitive erythrocyte differentiation ventral spinal cord interneuron differentiation cell differentiation in hindbrain commitment of neuronal cell to specific neuron type in forebrain central nervous system neuron development pituitary gland development response to lipid somatic stem cell population maintenance regulation of histone acetylation eosinophil fate commitment inner ear morphogenesis mast cell degranulation negative regulation of fat cell differentiation positive regulation of erythrocyte differentiation negative regulation of macrophage differentiation positive regulation of megakaryocyte differentiation positive regulation of neuron differentiation negative regulation of Notch signaling pathway positive regulation of angiogenesis positive regulation of transcription from RNA polymerase II promoter cell maturation digestive tract development homeostasis of number of cells within a tissue positive regulation of phagocytosis positive regulation of phagocytosis, engulfment definitive hemopoiesis semicircular canal development negative regulation of fat cell proliferation GABAergic neuron differentiation negative regulation of neural precursor cell proliferation regulation of forebrain neuron differentiation cell differentiation neuron differentiation negative regulation of myeloid cell differentiation neuron fate commitment
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7988999796-pwxjw:80/100.66.76.243:80.
          git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.39.1-2025.11.99-1.0