Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:2,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Recombinant Monoclonal |
Type |
Antibody |
Clone |
ARC1209 |
Immunogen |
A synthesized peptide derived from human GATA2/3 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1.8 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.3, with 50% glycerol, 0.05% BSA |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2849537 |
The genes for all 4 subunits of the T-cell antigen receptor (alpha, beta, gamma and delta) are controlled by distinct enhancers and their enhancer-binding proteins. Marine and Winoto (1991) identified a common TCR regulatory element by demonstrating binding of the enhancer-binding protein GATA3 to the enhancer elements of all 4 TCR genes. GATA3 had been shown in the chicken to be an enhancer-binding protein containing a zinc finger domain. GATA3 mRNA was demonstrated by Northern blot analysis in T cells but not in B cells or macrophages. GATA3 is abundantly expressed in the T-lymphocyte lineage and is thought to participate in T-cell receptor gene activation through binding to enhancers. Labastie et al. (1994) cloned the human gene and the 5-prime end of the mouse gene. The human gene comprises 6 exons distributed over 17 kb of DNA. Its 2 zinc fingers are encoded by 2 separate exons highly conserved with those of GATA1, but no other structural homologies between the 2 genes could be found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endothelial transcription factor GATA-2; GATA-binding factor 3; GATA-binding protein 2; GATA-binding protein 3; GATA-bindning protein 2; gata2 {ECO:0000312|EMBL:AAK51128.1}; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3
Gene Aliases: DCML; Gata-2; Gata-3; GATA2; GATA3; HDR; HDRS; IMD21; jal; MONOMAC; NFE1B
UniProt ID: (Human) P23769, (Human) P23771, (Mouse) O09100, (Rat) Q924Y4, (Mouse) P23772
Entrez Gene ID: (Human) 2624, (Human) 2625, (Mouse) 14461, (Rat) 25159, (Mouse) 14462, (Rat) 85471
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support