Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:1,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:100 | - |
Immunocytochemistry (ICC/IF) |
1:100-1:500 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
A synthetic peptide derived from the internal region of human GCNT7 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
Dulbecco's PBS, pH 7.4, with 50% glycerol, 150mM NaCl |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2555644 |
GCNT7, is a 430 amino acid glycosyltransferase that is localized to the Golgi apparatus. Other members of this family include GCNT1, GCNT2, GCNT3, GCNT4 and GCNT6. GCNT1 has been shown to play an important regulatory role in the biosynthesis of mucin-type O-glycans, which serve as ligands in cell adhesion. Specifically, GCNT1 expression in leukocytes regulates the synthesis of core 2 O-glycans that carry sialyl-Lewis x (sLex) oligosaccharides, which confer high affinity binding to Selectin proteins. Since downregulation of Selectin ligand expression has been shown to inhibit tissue infiltration, glycosyltransferase 14 family members represent potential drug targets for the treatment of inflammatory disorders and other pathologies involving Selectin proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 1,6-N-acetylglucosaminyltransferase; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 7
Gene Aliases: C20orf105; dJ1153D9.2; gcnt; GCNT7
UniProt ID: (Human) Q6ZNI0
Entrez Gene ID: (Human) 140687
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support