Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • GDF5 Antibodies

          Proteintech

          GDF5 Recombinant Rabbit Monoclonal Antibody (240944E5), CoraLite® Plus 488

          View all (12) GDF5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GDF5 Recombinant Rabbit Monoclonal Antibody (240944E5), CoraLite® Plus 488

          • Antibody Testing Data (1)
          GDF5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GDF5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GDF5 Antibody (CL488-83988-4) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed RAW 264.7 cells using CoraLite Plus 488 GDF5 antibody (CL488-83988-4, Clone: 240944E5 ) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GDF5 Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          CL488-83988-4

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240944E5

          Immunogen

          GDF5 fusion protein Ag6238

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Concentration

          1000 µg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Sequence of this protein is as follows: SFLLKKAREP GPPREPKEPF RPPPITPHEY MLSLYRTLSD ADRKGGNSSV KLEAGLANTI TSFIDKGQDD RGPVVRKQRY VFDISALEKD GLLGAELRIL RKKPSDTAKP AAPGGGRAAQ LKLSSCPSGR QPASLLDVRS VPGLDGSGWE VFDIWKLFRN FKNSAQLCLE LEAWERGRAV DLRGLGFDRA ARQVHEKALF LVFGRTKKRD LFFNEIKARS GQDDKTVYEY LFSQRRKRRA PLATRQGKRP SKNLKARCSR KALHVNFKDM GWDDWIIAPL EYEAFHCEGL CEFPLRSHLE PTNHAVIQTL MNSMDPESTP PTCCVPTRLS PISILFIDSA NNVVYKQYED MVVESCGCR

          Target Information

          GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BMP-14; Bone morphogenetic protein 14; brachypodism; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP-1; GDF-5; Growth/differentiation factor 5; H-GDF-5; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; Radotermin

          View more View less

          Gene Aliases: BDA1C; BMP-14; BMP14; Bp; brp; Cdmp-1; CDMP1; Gdf-5; GDF5; LAP-4; LAP4; OS5; SYM1B; SYNS2

          View more View less

          UniProt ID: (Human) P43026, (Mouse) P43027

          View more View less

          Entrez Gene ID: (Human) 8200, (Mouse) 14563

          View more View less

          Function(s)
          cytokine activity transforming growth factor beta receptor binding protein binding growth factor activity BMP binding identical protein binding receptor binding growth factor
          Process(es)
          chondrocyte differentiation transforming growth factor beta receptor signaling pathway SMAD protein import into nucleus cell-cell signaling positive regulation of pathway-restricted SMAD protein phosphorylation embryonic limb morphogenesis positive regulation of BMP signaling pathway negative regulation of chondrocyte differentiation positive regulation of chondrocyte differentiation forelimb morphogenesis hindlimb morphogenesis growth regulation of multicellular organism growth regulation of apoptotic process regulation of MAPK cascade negative regulation of neuron apoptotic process positive regulation of neuron differentiation cell development negative regulation of epithelial cell proliferation SMAD protein signal transduction chondroblast differentiation mitophagy in response to mitochondrial depolarization negative regulation of mesenchymal cell apoptotic process transmembrane receptor protein serine/threonine kinase signaling pathway BMP signaling pathway cartilage development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0