Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTSEFFSAQL RAQISDDTTH PISYYKPEFY MPDDGGTAHL SVVAEDGSAV SATSTINLYF GSKVRSPVSG ILLNNEMDDF SSTSITNEFG VPPSPANFIQ PGKQPLSSMC PTIMVGQDGQ VRMVVGAAGG TQITMATALA IIYNLWFGYD VKWAVEEPRL HNQLLPNVTT VERNIDQEVT AALETRHHHT QITSTFIAVV QAIVRMAGGW AAASDSRKGG EPAGY
This gene encodes a member of the gamma-glutamyl transpeptidase (GGT) family, which are important in the metabolism of glutathione. The most ubiquitously expressed human GGT gene, GGT1, encodes a single transmembrane polypeptide that is post-translationally processed to form a heavy and a light chain. In contrast, the product of this gene only contains homology to the light chain region, and lacks a transmembrane domain. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma-glutamyl transpeptidase; Gamma-glutamyltransferase light chain 1; gamma-glutamyltransferase-like activity 3; Gamma-glutamyltransferase-like activity 4; Gamma-glutamyltransferase-like protein 6; Glutathione hydrolase light chain 1
Gene Aliases: dJ831C21.1; dJ831C21.2; GGTL6; GGTLA3; GGTLA4; GGTLC1
UniProt ID: (Human) Q9BX51
Entrez Gene ID: (Human) 92086
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support