Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunoprecipitation (IP) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 350 and 400 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
1 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS, pH 7.0 to 8.0 |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
May act cooperatively with GRB10 to regulate tyrosine kinase receptor signaling. May increase IGF1 receptor phosphorylation under IGF1 stimulation as well as phosphorylation of IRS1 and SHC1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GRB10-interacting GYF protein 1; GYF domain containing 1; PERQ amino acid rich, with GYF domain 1; postmeiotic segregation increased 2-like 12
Gene Aliases: GYF1; PERQ1
Entrez Gene ID: (Human) 64599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support