Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: HeLa, HepG2
Immunogen sequence: MGNGMCSRKQ KRIFQTLLLL TVVFGFLYGA MLYYELQTQL RKAEAVALKY QQHQESLSAQ LQVVYEHRSR LEKSLQKERL EHKKAKEDFL VYKLEAQETL
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 130 kDa golgi-localized phosphoprotein; cis Golgi-localized calcium-binding protein; GIMPc; Golgi integral membrane protein 4; Golgi integral membrane protein, cis; Golgi phosphoprotein 4; Golgi phosphoprotein of 130 kDa; Golgi-localized phosphoprotein of 130 kDa; type II Golgi membrane protein
Gene Aliases: GIMPC; GOLIM4; GOLPH4; GPP130; P138
UniProt ID: (Human) O00461
Entrez Gene ID: (Human) 27333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support