Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. Such receptor phosphorylation initiates beta-arrestin-mediated receptor desensitization, internalization, and signaling events leading to their down-regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-CP4; G protein-coupled receptor kinase 2, groucho gene related; G protein-coupled receptor kinase 2-like; G protein-coupled receptor kinase 4; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase 6; G protein-coupled receptor kinase GRK4; G protein-coupled receptor kinase GRK5; G protein-coupled receptor kinase GRK6; G-protein coupled receptor kinase 6; ITI1; Poly(rC)-binding protein 4; unnamed protein product
Gene Aliases: 1200003L19Rik; A830025H08Rik; AlphaCP-4; FP2025; GPRK2L; GPRK4; GPRK5; GPRK6; GRK; GRK4; GRK4a; GRK5; GRK6; IT11; Pcbp4
UniProt ID: (Human) P32298, (Human) P34947, (Human) P43250, (Mouse) O70291, (Rat) P70507, (Mouse) Q8VEB1, (Rat) Q62833, (Mouse) O70293, (Mouse) P57724
Entrez Gene ID: (Human) 2868, (Human) 2869, (Human) 2870, (Mouse) 14772, (Rat) 59077, (Mouse) 14773, (Rat) 59075, (Mouse) 26385, (Rat) 59076, (Mouse) 59092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support