Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Genetic-screened homeobox 2; genomic screened homeo box 2 protein; GS homeobox 2; Homeobox protein GSH-2
Gene Aliases: GSH2; GSX2
UniProt ID: (Human) Q9BZM3
Entrez Gene ID: (Human) 170825, (Rat) 364140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support