Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
JWA protein expression is affected by vitamin A and may be associated with the cytoskeleton. A protein similar to JWA in rats may play a role in the regulation of cell differentiation by inhibiting the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor GTPase 6 interacting protein 5; ADP-ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor-like 6 interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation-like factor 6 interacting protein 5; aip-5; ARL-6-interacting protein 5; cytoskeleton related vitamin A responsive protein; Cytoskeleton-related vitamin A-responsive protein; dermal papilla derived protein 11; Dermal papilla-derived protein 11; Glutamate transporter EAAC1-interacting protein; glutamate transporter EEAC1-associated protein; GTRAP3-18; JM5; PRA1 domain family 3; PRA1 family protein 3; Prenylated Rab acceptor protein 2; Protein JWa; putative MAPK activating protein PM27; Putative MAPK-activating protein PM27
Gene Aliases: addicsin; ARL6IP5; DERP11; GTRAP3-18; hp22; HSPC127; jmx; JWA; PRA2; PRAF3; Yip6b
UniProt ID: (Human) O75915
Entrez Gene ID: (Human) 10550
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support