Product References
Galectin-3 and HBME-1 improve the accuracy of core biopsy in indeterminate thyroid nodules.
Endocrine
Trimboli P,Guidobaldi L,Amendola S,Nasrollah N,Romanelli F,Attanasio D,Ramacciato G,Saggiorato E,Valabrega S,Crescenzi A
MA5-12367 was used in immunohistochemistry - paraffin section to investigate using additional markers for core needle biopsy of thyroid nodules.
Fri Apr 01 00:00:00 EDT 2016
Higher bone marrow LGALS3 expression is an independent unfavorable prognostic factor for overall survival in patients with acute myeloid leukemia.
Blood
Cheng CL,Hou HA,Lee MC,Liu CY,Jhuang JY,Lai YJ,Lin CW,Chen HY,Liu FT,Chou WC,Chen CY,Tang JL,Yao M,Huang SY,Ko BS,Wu SJ,Tsay W,Tien HF
MA5-12367 was used in immunohistochemistry to study the prognostic value of bone marrow LGALS3 expression in acute myeloid leukemia
Thu Apr 18 00:00:00 EDT 2013
Immunohistochemical clue for the histological overlap of salivary adenoid cystic carcinoma and polymorphous low-grade adenocarcinoma.
Interventional medicine & applied science
El-Nagdy S,Salama NM,Mourad MI
MA5-12367 was used in immunohistochemistry to study the potential of immunohistochemistry to distinguish alivary adenoid cystic carcinoma and polymorphous low-grade adenocarcinoma
Sun Sep 01 00:00:00 EDT 2013
Investigation of galectin-3 and heparanase in endometrioid and serous carcinomas of the endometrium and correlation with known predictors of survival.
Archives of gynecology and obstetrics
Ege CB,Akbulut M,Zekioğlu O,Ozdemir N
MA5-12367 was used in immunohistochemistry to investigate the effect of galectin-3 and heparanase on the pathogenesis of endometrial carcinomas
Tue Nov 01 00:00:00 EDT 2011
Diagnostic and prognostic role of galectin 3 expression in cutaneous melanoma.
The American Journal of dermatopathology
Abdou AG,Hammam MA,Farargy SE,Farag AG,El Shafey EN,Farouk S,Elnaidany NF
MA5-12367 was used in immunohistochemistry to investigate the role of galectin 3 in the diagnosis and prognosis of cutaneous melanoma
Wed Dec 01 00:00:00 EST 2010
Immunohistochemical galectin-3 expression in non-melanoma skin cancers.
Pathology, research and practice
Kapucuoglu N,Basak PY,Bircan S,Sert S,Akkaya VB
MA5-12367 was used in immunohistochemistry to investigate the distribution of galectin-3 in cutaneous squamous cell carcinoma basal cell carcinoma
Tue May 19 00:00:00 EDT 2009
T cell apoptosis at the maternal-fetal interface in early human pregnancy, involvement of galectin-1.
Proceedings of the National Academy of Sciences of the United States of America
Kopcow HD,Rosetti F,Leung Y,Allan DS,Kutok JL,Strominger JL
MA5-12367 was used in western blot to investigate the effect of galectin-1 on the development of an immune-privileged environment at the maternal-fetal junction
Tue Nov 25 00:00:00 EST 2008
Galectin-3 expression in tumor progression and metastasis of papillary thyroid carcinoma.
Endocrine pathology
Türköz HK,Oksüz H,Yurdakul Z,Ozcan D
MA5-12367 was used in immunohistochemistry to examine the role of galectin-3 in the tumorigenesis of papillary thyroid carcinoma
Fri Oct 31 00:00:00 EDT 2008
Identification of immunohistochemical biomarkers for papillary thyroid carcinoma using gene expression profiling.
Human pathology
Murphy KM,Chen F,Clark DP
MA5-12367 was used in immunohistochemistry to identify the diagnostic markers for papillary thyroid carcinoma
Sat Mar 01 00:00:00 EST 2008
Galectin-3 expression is a potent prognostic marker in colorectal cancer.
Anticancer research
Endo K,Kohnoe S,Tsujita E,Watanabe A,Nakashima H,Baba H,Maehara Y
MA5-12367 was used in immunohistochemistry to investigate the prognostic significance of galectin-3 in colorectal cancer
Tue Sep 20 00:00:00 EDT 2005