Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAYVPAPGY QPTYNPTLPY YQPIPGGLNV GMSVYIQGVA SEHMKRFFVN FVVGQDPGSD VAFHFNPRFD GWDKVVFNTL QGGKWGSEER KRSMPFKKGA AFELVFIVLA EHYKVVVNGN PFYEYGHRLP LQMVTHLQVD GDLQLQSINF IGGQPLRPQG PPMMPPYPGP GHCHQQLNSL PTMEGPPTFN PPVPYFGRLQ GGLTARRTII IKGYVPPTGK SFAINFKVGS SGDIALHINP RMGNGTVVRN SLLNGSWGSE EKKITHNPFG PGQFFDLSIR CGLDRFKVYA NGQHLFDFAH RLSAFQRVDT LEIQGDVTLS YVQI (1-323 aa encoded by BC034750)
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. Galectin 4 binds lactose and a related range of sugars. May be involved in the assembly of adherens junctions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen NY-CO-27; Gal-4; Galectin-4; L-36 lactose-binding protein; L36LBP; Lactose-binding lectin 4; lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4
Gene Aliases: GAL4; L36LBP; LGALS4
UniProt ID: (Human) P56470
Entrez Gene ID: (Human) 3960
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support