Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
TXLNG gene ontology annotations related to this gene include regulation of bone mineralization; regulation of cell cycle process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; Factor inhibiting ATF4-mediated transcription; FIAT; Gamma-taxilin; lipopolysaccharide specific response-5 protein; Lipopolysaccharide-specific response protein 5
Gene Aliases: CXorf15; ELRG; FIAT; LSR5; TXLNG; TXLNGX
UniProt ID: (Human) Q9NUQ3
Entrez Gene ID: (Human) 55787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support