Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CTH Antibodies

          Proteintech

          Gamma cystathionase Monoclonal Antibody (2C7F9)

          24 References
          View all (32) CTH antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Gamma cystathionase Monoclonal Antibody (2C7F9)

          • Antibody Testing Data (15)
          Gamma cystathionase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Gamma cystathionase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Gamma cystathionase Antibody (60234-1-IG) in ICC/IF

          Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HepG2 cells using 60234-1-IG (Gamma cystathionase antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Mouse IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Gamma cystathionase Antibody in Immunocytochemistry (ICC/IF)
          Gamma cystathionase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gamma cystathionase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gamma cystathionase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gamma cystathionase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gamma cystathionase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gamma cystathionase Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Antibody in Western Blot (WB)
          Gamma cystathionase Monoclonal Antibody (2C7F9)

          Product Details

          60234-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:6,000
          View 18 publications 18 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:800
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 3 publications 3 publications

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Dog, Human, Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2C7F9

          Immunogen

          Gamma cystathionase Fusion Protein Ag2872 (74-406 aa encoded by BC015807)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: AVAALDGAK YCLAFASGLA ATVTITHLLK AGDQIICMDD VYGGTNRYFR QVASEFGLKI SFVDCSKIKL LEAAITPETK LVWIETPTNP TQKVIDIEGC AHIVHKHGDI ILVVDNTFMS PYFQRPLALG ADISMYSATK YMNGHSDVVM GLVSVNCESL HNRLRFLQNS LGAVPSPIDC YLCNRGLKTL HVRMEKHFKN GMAVAQFLES NPWVEKVIYP GLPSHPQHEL VKRQCTGCTG MVTFYIKGTL QHAEIFLKNL KLFTLAESLG GFESLAELPA IMTHASVLKN DRDVLGISDT LIRLSVGLED EEDLLEDLDQ ALKAAHPPSG SHS (74-406 aa encoded by BC015807)

          Target Information

          This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CGL; CTL target antigen; cystathionase (cystathionine gamma-lyase); Cystathionine gamma-lyase; Cysteine desulfhydrase; Cysteine-protein sulfhydrase; Gamma-cystathionase; Homocysteine desulfhydrase; homoserine deaminase; homoserine dehydratase; MGC9471; PRB-RA; Probasin-related antigen

          View more View less

          Gene Aliases: 0610010I13Rik; AI314617; BC019483; CSE; CTH; Cys3; H

          View more View less

          UniProt ID: (Human) P32929, (Rat) P18757, (Mouse) Q8VCN5

          View more View less

          Entrez Gene ID: (Human) 1491, (Rat) 24962, (Mouse) 107869

          View more View less

          Function(s)
          cystathionine gamma-synthase activity cystathionine gamma-lyase activity protein binding calmodulin binding carbon-sulfur lyase activity pyridoxal phosphate binding L-cystine L-cysteine-lyase (deaminating) homocysteine desulfhydrase activity L-cysteine desulfhydrase activity cystathionine beta-lyase activity catalytic activity lyase activity lyase
          Process(es)
          sulfur amino acid catabolic process cysteine metabolic process protein-pyridoxal-5-phosphate linkage via peptidyl-N6-pyridoxal phosphate-L-lysine cysteine biosynthetic process via cystathionine cysteine biosynthetic process transsulfuration endoplasmic reticulum unfolded protein response positive regulation of I-kappaB kinase/NF-kappaB signaling protein sulfhydration positive regulation of NF-kappaB transcription factor activity protein homotetramerization hydrogen sulfide biosynthetic process 'de novo' L-methionine biosynthetic process positive regulation of aortic smooth muscle cell differentiation negative regulation of apoptotic signaling pathway glutathione metabolic process negative regulation of cell proliferation negative regulation of cell growth negative regulation of apoptotic process homocysteine metabolic process cellular amino acid biosynthetic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0