Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gephyrin; gephyrin isoform; Putative glycine receptor-tubulin linker protein
Gene Aliases: 5730552E08Rik; AI662856; BC027112; C230040D23; GEPH; GPH; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC
UniProt ID: (Human) Q9NQX3, (Rat) Q03555, (Mouse) Q8BUV3
Entrez Gene ID: (Human) 10243, (Rat) 64845, (Mouse) 268566
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support