Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NMDAR2B Antibodies

          Antibodies Inc

          GluN2B/NR2B Glutamate Receptor Monoclonal Antibody (N59/36), FL550

          View all (66) NMDAR2B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GluN2B/NR2B Glutamate Receptor Monoclonal Antibody (N59/36), FL550

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          GluN2B/NR2B Glutamate Receptor Monoclonal Antibody (N59/36), FL550

          Product Details

          75-101-FL550

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N59/36

          Immunogen

          Fusion protein amino acids 20-271 (extracellular N-terminus) of rat NR2B (accession number Q00960) produced recombinantly in E. Coli.
          View immunogen

          Conjugate

          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation

          Excitation/Emission Max

          550/575 nm

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Dry ice (international)

          Product Specific Information

          For maximum recovery of product, centrifuge the vial prior to removing the cap.

          Target Information

          NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer's Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal dominant 6-related mental retardation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GluN2B; GluN2B(alt_5'UTR); GluRepsilon2; Glutamate [NMDA] receptor subunit epsilon-2; glutamate receptor; Glutamate receptor ionotropic, NMDA 2B; glutamate receptor subunit epsilon-2; glutamate receptor, ionotropic, N-methyl D-aspartate 2B; glutamate receptor, ionotropic, NMDA2B; GRIN 2B; HGNC:4586; hNR 3; MGC142178; MGC142180; N-methyl D-aspartate receptor 2B; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 3; NMDAR2B; NMDE2; NME2; NR2B; NR3

          View more View less

          Gene Aliases: AW490526; DEE27; EIEE27; GluN2B; GRIN2B; hNR3; MRD6; NMDAR2B; NR2B; NR3

          View more View less

          UniProt ID: (Human) Q13224, (Mouse) Q01097, (Rat) Q00960

          View more View less

          Entrez Gene ID: (Human) 2904, (Mouse) 14812, (Rat) 24410

          View more View less

          Function(s)
          beta-amyloid binding NMDA glutamate receptor activity ion channel activity protein binding zinc ion binding ligand-gated ion channel activity glycine binding glutamate binding glutamate-gated calcium ion channel activity signaling receptor activity metal ion binding ligand-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential receptor activity ionotropic glutamate receptor activity receptor binding interleukin-1 receptor binding extracellular-glutamate-gated ion channel activity cation channel activity calcium channel activity beta-catenin binding drug binding D2 dopamine receptor binding ionotropic glutamate receptor binding neurotransmitter binding protein heterodimerization activity cell adhesion molecule binding transmembrane signal receptor
          Process(es)
          ion transport glutamate receptor signaling pathway chemical synaptic transmission brain development learning or memory calcium-mediated signaling ion transmembrane transport ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic regulation of membrane potential response to ethanol regulation of synaptic plasticity regulation of neuronal synaptic plasticity protein heterotetramerization positive regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential long-term synaptic potentiation calcium ion transmembrane import into cytosol cation transmembrane transport excitatory chemical synaptic transmission regulation of presynaptic membrane potential negative regulation of dendritic spine maintenance regulation of cation transmembrane transport positive regulation of excitatory postsynaptic potential action potential behavioral fear response in utero embryonic development startle response suckling behavior transport cation transport calcium ion transport sensory organ development learning memory associative learning embryo development regulation of protein kinase A signaling positive regulation of cell death positive regulation of glutamate secretion fear response receptor clustering regulation of MAPK cascade regulation of long-term neuronal synaptic plasticity behavioral response to pain rhythmic process positive regulation of synaptic transmission detection of mechanical stimulus involved in sensory perception of pain regulation of postsynaptic membrane potential calcium ion transmembrane transport response to amphetamine long-term memory response to mechanical stimulus response to toxic substance response to carbohydrate response to manganese ion cellular response to magnesium starvation response to organic cyclic compound response to amine hippocampus development cerebral cortex development response to magnesium ion multicellular organismal response to stress response to cytokine response to cocaine response to calcium ion response to methylmercury response to electrical stimulus response to other organism response to growth hormone response to fungicide cellular response to amino acid stimulus cellular response to manganese ion cellular response to dsRNA cellular response to growth factor stimulus cellular response to lipid cellular response to organic cyclic compound
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0