Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:3,000 | - |
ELISA (ELISA) |
1:5,000-1:25,000 | - |
Immunoprecipitation (IP) |
1:500-1:3,000 | - |
Product Specifications | |
---|---|
Species Reactivity |
Bacteria |
Host/Isotype |
Goat / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Glucose-6-Phosphate Dehydrogenase [Leuconostoc mesenteroides] |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
1 mg/mL |
Purification |
Ion-exchange chromatography |
Storage buffer |
0.02M potassium phosphate, pH 7.2, with 0.15M NaCl |
Contains |
0.01% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-GLUCOSE-6-PHOSPHATE DEHYDROGENASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified Glucose-6-Phosphate Dehydrogenase [Leuconostoc mesenteroides]. Cross reactivity against Glucose-6-Phosphate Dehydrogenase from other tissues and species may occur but have not been specifically determined.
This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support