Invitrogen
Applications | Tested Dilution |
---|---|
Immunocytochemistry (ICC/IF) |
1-10 µg/mL |
Product Specifications | |
---|---|
Species Reactivity |
Chicken |
Host/Isotype |
Goat / IgG |
Class |
Polyclonal |
Type |
Secondary Antibody |
Immunogen |
Gamma Immunoglobins Heavy and Light chains |
Conjugate |
DSB-X Biotin
|
Form |
liquid |
Concentration |
2 mg/mL |
Storage conditions |
4° C, store in dark |
RRID |
AB_2536520 |
Target |
IgY |
Antibody Form |
Whole Antibody |
The DSB-X biotin goat anti-chicken IgG is prepared from antibodies that have not been cross-adsorbed. DSB-X biotin is a derivative of desthiobiotin, a stable biotin percursuor. DSB-X biotin utilizes a seven-atom spacer to increase the available of the DSB-Xbiotin conjugate to bind in the deep biotin-binding pocket of streptpavidin or avidin. Whereas harsh chaotropic agents and low pH are required to dissociate a biotin complex from avidin or steptavidin, binding can be reversed by adding excess D-desthiobiotin or natural D-biotin at neutral pH and at room temperature.
Obtain superior images with the new Invitrogen™ Alexa Fluor™ Plus secondary antibodies. Make your low abundant targets visible, minimize time spent optimizing, and make your precious samples count. Alexa Fluor™ Plus secondary antibodies provide brighter signal while having lower cross-reactivity compared to leading Alexa Fluor secondary antibodies, providing you with higher sensitivity and better signal-to-noise for your critical experiments. Alexa Fluor Plus secondary antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor Plus secondary antibodies are conjugated using new, proprietary dye chemistry and are pre-adsorbed to reduce background and cross-reactivity so you can generate stunning images. Alexa Fluor Plus secondary antibodies provide up to 4.2 fold higher signal-to-background in imaging applications and up to 5.8 fold higher signal-to-background in fluorescent western applications compared to Alexa Fluor secondary conjugates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support