Invitrogen
Catalog #:A28175SAMPLE
Quantity:
Usually Ships In 1 business day
| TESTED APPLICATIONS | DILUTION |
|---|---|
| Flow Cytometry (Flow) | 1-5 µg/ml |
| Immunocytochemistry (ICC) | 0.1-2 µg/ml |
| Immunofluorescence (IF) | 0.1-2 µg/ml |
| Tested Species reactivity | Mouse |
| Host / Isotype | Goat / IgG |
| Class | Oligoclonal |
| Type | Secondary Antibody |
| Immunogen | Recombinant full-length protein |
| Conjugate | Alexa Fluor® 488 |
| Excitation/Emission Profile | View spectra |
| Form | Liquid |
| Concentration | 1 mg/ml |
| Purification | Gravity column chromatography |
| Storage buffer | PBS |
| Contains | 5mM sodium azide |
| Storage conditions | 4° C, store in dark, DO NOT FREEZE! |
| RRID | AB_2610666 |
| Target | IgG |
| Antibody Form | Whole Antibody |
This is a sample size of part number A28175.
The sensitivity and specificity of each lot is confirmed using ELISA.
Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig and donkey IgG is observed.
Superclonal™ Secondary Antibodies are developed to be highly specific for both heavy and light chains of IgG. Utilizing the power of recombinant technology and multiple phenotypic screens, Superclonal™ antibodies are selected for enhanced sensitivity and specificity to the target species IgG. These recombinant secondary antibodies show exceptional performance in common immunoassay applications, with very little cross-reactivity to other commonly-used species. In addition, their recombinant animal-free-origin ensures lot-to-lot consistency, hence eliminating the need to optimize each lot in relevant assays. Our Superclonal™ secondary antibodies allow scientists to achieve the accurate results they need, without the cross-reactivity concerns that can impact some traditional polyclonal secondary antibodies. Superclonal™ secondaries are available with Alexa Fluor conjugates, as well as HRP, AP and biotin. The Alexa Fluor dyes have set the industry standard for brightness and photostability for several years. Alexa Fluor secondary antibodies are trusted, having been used in over 30,000 peer reviewed papers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.