Invitrogen
Applications | Tested Dilution |
---|---|
Immunocytochemistry (ICC/IF) |
1 µg/mL |
Product Specifications | |
---|---|
Species Reactivity |
Rabbit |
Host/Isotype |
Goat / IgG |
Class |
Polyclonal |
Type |
Secondary Antibody |
Immunogen |
Gamma Immunoglobins Heavy and Light chains |
Conjugate |
FITC
|
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Purification |
purified |
Storage buffer |
PBS, pH 7.2, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
RRID |
AB_2536006 |
Target |
IgG |
Cross Adsorption |
Against bovine, human and mouse IgG and serum proteins |
Antibody Form |
F(ab')2 Fragment |
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support