Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Applications | Tested Dilution |
---|---|
Western Blot (WB) |
1:1,000-1:20,000 |
Immunohistochemistry (IHC) |
1:500-1:20,000 |
ELISA (ELISA) |
1:500-1:20,000 |
Product Specifications | |
---|---|
Species Reactivity |
Rat |
Host/Isotype |
Goat / IgG |
Class |
Polyclonal |
Type |
Secondary Antibody |
Immunogen |
Gamma Immunoglobins Heavy and Light chains |
Conjugate |
HRP
|
Form |
Lyophilized |
Concentration |
1 mg/mL |
Purification |
Affinity Chromatography |
Storage buffer |
PBS, pH 7.2, with 1% BSA |
Contains |
0.1% ProClin 150 |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2536017 |
Target |
IgG |
Cross Adsorption |
Against human and mouse IgG |
Antibody Form |
F(ab')2 Fragment |
Rehydrate with 0.55 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin rat serum proteins, IgG from human or mouse, highly cross absorbed against mouse IgG.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support