Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1/50 - 1/500 | View 1 publication 1 publication |
Immunohistochemistry (Paraffin) (IHC (P)) |
1/500 - 1/4000 | - |
Immunohistochemistry (Frozen) (IHC (F)) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Dog, Cat, Human, Sheep, Rat |
Published species |
Not Applicable |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Growth hormone from ovine pituitary. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
5 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS |
Contains |
0.09% sodium azide |
Storage conditions |
4°C or -20°C if preferred |
RRID |
AB_2111267 |
In Western blot applications a band at ~20 kDa is seen in human pituitary gland lysates.
Rabbit anti growth hormone antibody recognizes growth hormone (GH), which acts to control body growth and metabolic activity.
Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GH; Growth hormone; Growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; HGH; Pituitary growth hormone; Somatotropin
Gene Aliases: GH; GH-N; GH1; GHB1; GHB5; GHN; hGH-N; IGHD1B; RNGHGP
UniProt ID: (Sheep) P67930, (Dog) P33711, (Cat) P46404, (Human) P01241, (Mouse) P06880, (Rat) P01244
Entrez Gene ID: (Sheep) 443329, (Dog) 403795, (Cat) 493931, (Human) 2688, (Mouse) 14599, (Rat) 24391
Molecular Function:
peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support