Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-18005 targets H-2Ld in FACS applications and shows reactivity with Mouse samples.
The MA5-18005 immunogen is mouse H-2Ld (public determinant H-2.65).
MHC Class I (H-2Ld) is a protein complex located on the surface of cells in vertebrates. Its main function is to present peptides derived from intracellular proteins to T cells for immune recognition. The structure of MHC Class I (H-2Ld) consists of a heavy chain, beta-2 microglobulin, and a peptide-binding groove that interacts with antigenic peptides. The location of MHC Class I (H-2Ld) varies depending on the cell type, but it is typically found on the surface of nucleated cells.
HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H-2L; H2L; H2LD; histocompatibility 2, D region locus L; histocompatibility 2, L region
Gene Aliases: H-2L
Entrez Gene ID: (Mouse) 14980
Molecular Function: major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support