Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-Histone H3 [Trimethyl Lys18] reacts with human Histone H3.2. A BLAST analysis was used to suggest cross-reactivity with Human, mouse, and C. elegans. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. Cross-reactivity with Histone H3 from other sources has not been determined.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in a histone cluster on chromosome 1. This gene is one of four histone genes in the cluster that are duplicated; this record represents the telomeric copy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG5825-PA; CG5825-PC; CG5825-PD; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3B (H3.3B); H3-clustered histone 13; H3-clustered histone 14; H3-clustered histone 15; H3-clustered histone 2; H3-clustered histone 3; H3-clustered histone 4; H3-clustered histone 6; H3-clustered histone 7; H3.A; H3.B; H3K56me3; H3L-like histone; H3Lys18me3; H3Lys56me3; His3.3A-PA; His3.3A-PC; His3.3A-PD; histone ; histone 1, H3b; histone 2, H3a; histone 2, H3c; histone 2, H3c2; histone 2, H3ca2; histone cluster 2, H3c2, pseudogene; Histone H2A; Histone H3.1; Histone H3.2; Histone H3.3; histone H3.3 ; Histone H3.3A; Histone H3.3B; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l; Histone H3/m; Histone H3/o; Tri-methyl H3; Tri-methyl Histone H3; Trimethyl H3; Trimethyl Histone H3
Gene Aliases: B0035.7; CELE_T10C6.12; CG5825; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; H3; H3-143; H3-3A; H3-3B; H3-53; H3-614; H3-B; H3-F; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; H3.3; H3.3A; H3.3B; H3/l; H3/M; H3/n; H3/o; H3b; H3C1; H3C10; H3C11; H3C12; H3C13; H3C14; H3C15; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3f; H3F2; H3F3; H3F3A; H3F3B; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; his-12; his-16; his-19; his-21; his-3; His-3.3A; his-30; his-33; his-43; his-47; his-51; his-53; his-57; his-61; his-65; his-68; his-7; His3.3; His3.3A; His3.3B; HIST1H3A; HIST1H3B; Hist1h3c; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; HIST2H3A; Hist2h3b; HIST2H3C; Hist2h3c1; Hist2h3c2; Hist2h3c2-ps; Hist2h3ca1; Hist2h3ca2; HIST2H3D; K06C4.11; K06C4.3; M32461; PH3; PP781; T10C6.12; T23D8.6; ZK131.10; ZK131.6
UniProt ID: (Human) P84243, (Human) P68431, (Human) Q71DI3, (Fruit fly) C0HL66, (Mouse) P84228, (C. elegans) P09588
Entrez Gene ID: (Human) 3021, (Human) 8358, (Human) 333932, (Human) 126961, (Fruit fly) 33736, (Mouse) 319150, (Mouse) 97114, (C. elegans) 180073
Molecular Function: histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support