Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: Human coronavirus (HCoV-HKU1) Nucleocapsid Protein (His Tag).
Immunogen sequence: WYRHSRRSFK TADGQQKQLL PRWYFYYLGT GPYANASYGE SLEGVFWVAN HQADTSTPSD VSSRDPTTQE AIPTRFPPGT ILPQGYYVEG SGRSASNSRP G
Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP) and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M. Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coronavirus; HCoV; HKU1; Nucleoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support