Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1-5 µg/mL | - |
ELISA (ELISA) |
0.1 ng/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG2a, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
2F10 |
Immunogen |
HEY1 (NP_036390, 121 a.a. approximately 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
See Label |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.4 |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
Sequence of this protein is as follows: DYRSLGFREC LAEVARYLSI IEGLDASDPL RVRLVSHLNN YASQREAASG AHAGLGHIPW GTVFGHHPHI AHPLLLPQNG HGNAGTTASP TEPHHQGRLG
Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic helix-loop-helix protein OAF1; bHLHb31; Cardiovascular helix-loop-helix factor 2; CHF-2; Class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy-related transcription factor 1; hairy/enhancer-of-split related with YRPW motif 1; Hairy/enhancer-of-split related with YRPW motif protein 1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; HESR-1; HRT-1; MGC1274
Gene Aliases: BHLHB31; CHF2; HERP2; HESR1; HEY1; hHRT1; HRT-1; HRT1; OAF1
UniProt ID: (Human) Q9Y5J3
Entrez Gene ID: (Human) 23462
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support