Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HEY2 Antibodies

          Proteintech

          HEY2 Recombinant Rabbit Monoclonal Antibody (240474F2)

          View all (13) HEY2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HEY2 Recombinant Rabbit Monoclonal Antibody (240474F2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          HEY2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HEY2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HEY2 Antibody (83501-7-RR) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using HEY2 antibody (83501-7-RR, Clone: 240474F2) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HEY2 Antibody in Immunocytochemistry (ICC/IF)
          HEY2 Antibody in Immunocytochemistry (ICC/IF)
          HEY2 Antibody in Flow Cytometry (Flow)
          HEY2 Antibody in Flow Cytometry (Flow)
          HEY2 Antibody in Surface plasmon resonance (SPR)
          HEY2 Recombinant Rabbit Monoclonal Antibody (240474F2)

          Product Details

          83501-7-RR

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:200-1:800
          -

          Flow Cytometry (Flow)

          0.25 µg/10^6 cells in a 100 µL suspension
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Surface plasmon resonance (SPR)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293F

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          240474F2

          Immunogen

          HEY2 fusion protein Ag30988
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MKRPCEETTS ESDMDETIDV GSENNYSGQS TSSVIRLNSP TTTSQIMARK KRRGIIEKRR RDRINNSLSE LRRLVPTAFE KQGSAKLEKA EILQMTVDHL KMLQATGGKG YFDAHALAMD FMSIGFRECL TEVARYLSSV EGLDSSDPLR VRLVSHLSTC ATQREAAAMT SSMAHHHHPL HPHHWAAAFH HLPAALLQPN GLHASESTPC RLSTTSEVPP AHGSALLTAT FAHADSALRM PSTGSVAPCV PPLSTSLLSL SATVHAAAAA ATAAAHSFPL SFAGAFPMLP PNAAAAVAAA TAISPPLSVS ATSSPQQTSS GTNNKPYRPW GTEVGAF

          Target Information

          Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bHLHb32; cardiovascular basic helix-loop-helix factor 1; Cardiovascular helix-loop-helix factor 1; CHF1; Class B basic helix-loop-helix protein 32; h hHRT2; Hairy and enhancer of split-related protein 2; Hairy-related transcription factor 2; hairy/enhancer-of-split related with YRPW motif 2; Hairy/enhancer-of-split related with YRPW motif protein 2; hCHF1; HES-related repressor protein 1; HES-related repressor protein 2; HESR-2; hGRL; basic helix-loop-helix transcription factor; hHRT2; HRT-2; HRT2; Protein gridlock homolog; transcription factor; unnamed protein product

          View more View less

          Gene Aliases: BHLHB32; CHF1; GRIDLOCK; GRL; HERP; HERP1; HESR2; HEY2; HRT2

          View more View less

          UniProt ID: (Human) Q9UBP5

          View more View less

          Entrez Gene ID: (Human) 23493

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding identical protein binding histone deacetylase binding sequence-specific DNA binding protein dimerization activity RNA polymerase II sequence-specific DNA binding transcription factor binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter blood vessel development vasculogenesis muscular septum morphogenesis outflow tract morphogenesis cardiac conduction system development atrioventricular valve development aortic valve morphogenesis pulmonary valve morphogenesis tricuspid valve morphogenesis tricuspid valve formation epithelial to mesenchymal transition involved in endocardial cushion formation endocardial cushion to mesenchymal transition involved in heart valve formation cardiac ventricle morphogenesis cardiac left ventricle morphogenesis cardiac right ventricle morphogenesis ventricular trabecula myocardium morphogenesis cardiac muscle hypertrophy regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter Notch signaling pathway pattern specification process heart development anatomical structure morphogenesis anterior/posterior axis specification anterior/posterior pattern specification positive regulation of heart rate regulation of gene expression negative regulation of gene expression cardiac muscle cell apoptotic process negative regulation of cardiac muscle cell apoptotic process mesenchymal cell development cardiac muscle hypertrophy in response to stress ascending aorta morphogenesis dorsal aorta morphogenesis umbilical cord morphogenesis cell fate commitment regulation of auditory receptor cell differentiation negative regulation of Notch signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter regulation of neurogenesis smooth muscle cell differentiation negative regulation of smooth muscle cell differentiation ventricular cardiac muscle cell development cardiac muscle cell proliferation positive regulation of cardiac muscle cell proliferation cardiac epithelial to mesenchymal transition heart trabecula formation cardiac septum morphogenesis ventricular septum morphogenesis atrial septum morphogenesis negative regulation of transcription initiation from RNA polymerase II promoter labyrinthine layer blood vessel development blood vessel endothelial cell differentiation artery development arterial endothelial cell differentiation cardiac vascular smooth muscle cell development coronary vasculature morphogenesis pulmonary artery morphogenesis protein-DNA complex assembly negative regulation of biomineral tissue development circulatory system development cochlea development vascular smooth muscle cell development negative regulation of cardiac vascular smooth muscle cell differentiation regulation of vasculogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline