Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: STSVTGQVLG GPHNLMRRST VSLLDTYQKC GLKRKSEEIE NTSSVQIIEE HPPMIQNNAS GATVATATTS TATSKNSGSN SEGDYQLVQH EVLCSMTNTY E
HIPK2 is a nuclear serine/threonine kinase that is linked to transcriptional regulation and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hHIPk2; Homeodomain-interacting protein kinase 2
Gene Aliases: HIPK2; PRO0593
UniProt ID: (Human) Q9H2X6
Entrez Gene ID: (Human) 28996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support