Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MDCEHSLSLP CRGAEAWEIG IDKYLYAMRL SDETLIDIMT RFRKEMKNGL SRDFNPTATV KMLPTFVRSI PDGSEKGDFI ALDLGGSSFR ILRVQVNHEK NQNVHMESEV YDTPENIVHG SGSQLFDHVA ECLGDFMEKR KIKDKKLPVG FTFSFPCQQS KIDEAILITW TKRFKASGVE GADVVKLLNK AIKKRGDYDA NIVAVVNDTV GTMMTCGYDD QHCEVGLIIG TGTNACYMEE LRHIDLVEGD EGRMCINTEW GAFGDDGSLE DIRTEFDREI DRGSLNPGKQ LFEKMVSGMY LGELVRLILV KMAKEGLLFE GRITPELLTR GKFNTSDVSA IEKNKEGLHN AKEILTRLGV EPSDDDCVSV QHVCTIVSFR SANLVAATLG AILNRLRDNK GTPRLRTTVG VDGSLYKTHP QYSRRFHKTL RRLVPDSDVR FLLSESGSGK GAAMVTAVAY RLAEQHRQIE ETLAHFHLTK DMLLEVKKRM RAEMELGLRK QTHNNAVVKM LPSFVRRTPD GTENGDFLAL DLGGTNFRVL LVKIRSGKKR TVEMHNKIYA IPIEIMQGTG EELFDHIVSC ISDFLDYMGI KGPRMPLGFT FSFPCQQTSL DAGILITWTK GFKATDCVGH DVVTLLRDAI KRREEFDLDV VAVVNDTVGT MMTCAYEEPT CEVGLIVGTG SNACYMEEMK NVEMVEGDQG QMCINMEWGA FGDNGCLDDI RTHYDRLVDE YSLNAGKQRY EKMISGMYLG EIVRNILIDF TKKGFLFRGQ ISETLKTRGI FETKFLSQIE SDRLALLQVR AILQQLGLNS TCDDSILVKT VCGVVSRRAA QLCGAGMAAV VDKIRENRGL DRLNVTVGVD GTLYKLHPHF SRIMHQTVKE LSPKCNVSFL LSEDGSGKGA ALITAVGVRL RTEASS
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain form hexokinase; glycolytic enzyme; hexokinase I; hexokinase IR; Hexokinase type I; Hexokinase-1; Hexokinase-A; HK I
Gene Aliases: HEXOKIN; hexokinase; HK; HK1; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1
UniProt ID: (Human) P19367, (Rat) P05708
Entrez Gene ID: (Human) 3098, (Rat) 25058
Molecular Function: kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support