Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product # 7KMWCO
HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b2 microglobulin; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-G histocompatibility antigen, class I, G; MHC class I antigen G; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G
Gene Aliases: HLA-6.0; HLA-G; HLAG; MHC-G
UniProt ID: (Human) P17693
Entrez Gene ID: (Human) 3135
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support