Proteintech
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000-1:10,000 | View 1 publication 1 publication |
Immunohistochemistry (IHC) |
- | View 1 publication 1 publication |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:1,000-1:10,000 | - |
Flow Cytometry (Flow) |
0.2 µg/1x10^6 cells | - |
Miscellaneous PubMed (Misc) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Human, Pig |
Published species |
Human |
Host/Isotype |
Mouse / IgG2b |
Class |
Monoclonal |
Type |
Antibody |
Clone |
5C5B7 |
Immunogen |
HLA class I ABC Fusion Protein Ag7488 (28-303 aa encoded by BC003069) |
Conjugate |
Unconjugated
|
Form |
Liquid |
Concentration |
1.11 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: SMRYFFTSV SRPGRGEPRF IAVGYVDDTQ FVRFDSDAAS QKMEPRAPWI EQEGPEYWDQ ETRNMKAHSQ TDRANLGTLR GYYNQSEDGS HTIQIMYGCD VGPDGRFLRG YRQDAYDGKD YIALNEDLRS WTAADMAAQI TKRKWEAVHA AEQRRVYLEG RCVDGLRRYL ENGKETLQRT DPPKTHMTHH PISDHEATLR CWALGFYPAE ITLTWQRDGE DQTQDTELVE TRPAGDGTFQ KWAAVVVPSG EEQRYTCHVQ HEGLPKPLTL RWELSSQ (28-303 aa encoded by BC003069)
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determinants important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumor defense. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A29 protein; alpha chain of MHC class II; antigen presenting molecule; class I MHC, HLA-B antigen; epididymis secretory protein Li 83; H-2K; H-2K(d); H2-K; histocompatibility antigen; HLA -B; HLA A; HLA B-40011; HLA B44; HLA C; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA class I histocompatibility antigen, C alpha chain; HLA class I histocompatibility antigen, Cw-15 alpha chain; HLA class I histocompatibility antigen, Cw-18 alpha chain; HLA locus; HLA molecule; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-Aw33.1; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B35; HLA-B35 antigen; HLA-Bw50 antigen; HLA-C; HLA-C alpha chain; HLA-C alpha chain (HLA-Cw*15v); HLA-C alpha chain for Cw*18GB; HLA-C alpha-chain; HLA-C antigen; HLA-Cw; HLA-Cw*04GB; HLA-Cw*1702; HLA-Cw*18GB; HLA-Cw-1503; Human leukocyte antigen A; Human leukocyte antigen B; Human leukocyte antigen C; human leukocyte antigen-C alpha chain; hypothetical protein; lecocyte antigen B; leucocyte antigen A; Leukocyte Antigen; leukocyte antigen C; leukocyte antigen class I-A; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility antigen HLA-C; MCH class I antigen; MGC184092; MGC7052; MHC 1; MHC antigen; MHC clas I antigen; MHC Class 1; MHC class 1 antigen; MHC class I; MHC class I alpha chain; MHC class I anitgen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen Cw*15; MHC class I antigen Cw*18; MHC class I antigen GN00104; MHC class I antigen heavy chain; MHC class I antigen heavy chain HLA-C; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen null protein; MHC class I antigen SHCHA; MHC class I antigene; MHC class I histocompatibility antigen; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-ABC; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27-HS; MHC HLA-B27d; MHC HLA-B39N; MHC I HLA; MHC1; surface antigen; truncated class I antigen; truncated MHC class I antigen
Gene Aliases: AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; D6S204; HEL-S-83; HLA-A; HLA-A11; HLA-A33; HLA-B; HLA-B*45ZJ; HLA-B-3506; HLA-B-3905; HLA-B-5502; HLA-B-5602; HLA-B15; HLA-B39; HLA-B49; HLA-B50; HLA-B55; HLA-B59; HLA-B61; HLA-C; HLA-Cw; HLA-DQB1; HLA-DRB1; HLA-JY3; HLAA; HLAB; HLAC; HLC-C; MHC; PSORS1; SPDA1
UniProt ID: (Human) P30444, (Human) P79523, (Human) P10321
Entrez Gene ID: (Human) 3105, (Human) 3106, (Human) 3107, (Pig) 100515902
Molecular Function:
major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support