Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:2,000-1:10,000 | - |
Immunoprecipitation (IP) |
2-10 µg/mg lysate | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 375 and 425 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
1 mg/mL |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
HLX is a transcription factor required for TBX21/T-bet-dependent maturation of Th1 cells as well as maintenance of Th1-specific gene expression. Involved in embryogenesis and hematopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H2.0-like homeo box 1; H2.0-like homeo box-1; H2.0-like homeobox 1; H2.0-like homeobox protein; Homeobox protein HB24; Homeobox protein HLX1
Gene Aliases: HLX; HLX1
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support