Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:500-1:2,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:200 | - |
Immunocytochemistry (ICC/IF) |
1:50-1:200 | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Recombinant full length Human HMG20A. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.2, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
RRID |
AB_2720916 |
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10739; High mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1; iBRAF; Inhibitor of BRAF35
Gene Aliases: 1200004E06Rik; 5730490E10Rik; HMG20A; HMGX1; HMGXB1; Ibraf; RGD1564760
UniProt ID: (Human) Q9NP66, (Mouse) Q9DC33
Entrez Gene ID: (Human) 10363, (Mouse) 66867, (Rat) 315689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support