Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGEQPIFSTR AHVFQIDPNT KKNWVPTSKH AVTVSYFYDS TRNVYRIISL DGSKAIINST ITPNMTFTKT SQKFGQWADS RANTVYGLGF SSEHHLSKFA EKFQEFKEAA RLAKEKSQEK MELTSTPSQE SAGGDLQSPL TPESINGTDD ERTPDVTQNS EPRAEPTQNA LPFSHSSAIS KHWEAELATL KGNNAKLTAA LLESTANVKQ WKQQLAAYQE EAERLHKRVT ELECVSSQAN AVHTHKTELN QTIQELEETL KLKEEEIERL KQEIDNAREL QEQRDSLTQK LQEVEIRNKD LEGQLSDLEQ RLEKSQNEQE AFRNNLKTLL EILDGKIFEL TELRDNLAKL LECS
HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homer 1; homer homolog 1; Homer protein homolog 1; homer scaffolding protein 1; homer, neuronal immediate early gene, 1; Homer-1; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; VASP/Ena-related gene up-regulated during seizure and LTP 1; Vesl-1
Gene Aliases: HOMER; homer-1; HOMER1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; PSD-Zip45; SYN47; Ves-1; Vesl; Vesl-1; Vesl1
UniProt ID: (Human) Q86YM7, (Mouse) Q9Z2Y3, (Rat) Q9Z214
Entrez Gene ID: (Chicken) 427181, (Rabbit) 100356113, (Human) 9456, (Pig) 100511070, (Mouse) 26556, (Rat) 29546
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support