HOMER1 is a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homer homolog 1; Homer protein homolog 1; homer, neuronal immediate early gene, 1; Homer-1; immediate early gene protein Homer1A; PSD-Zip45; scaffold protein; VASP/Ena-related gene up-regulated during seizure and LTP 1; Vesl-1
Gene Aliases: HOMER; homer-1; HOMER1; HOMER1A; HOMER1B; HOMER1C; HOMER1F; PSD-Zip45; SYN47; Ves-1; Vesl; Vesl-1; Vesl1
UniProt ID: (Human) Q86YM7, (Mouse) Q9Z2Y3, (Rat) Q9Z214
Entrez Gene ID: (Human) 9456, (Mouse) 26556, (Rat) 29546
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support