Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200 | - |
Immunohistochemistry (Frozen) (IHC (F)) |
1:200 | - |
Immunocytochemistry (ICC/IF) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide corresponding to amino acid residues 302-316 of rat Homer2 (Intracellular, C-terminus) |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.8 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2857303 |
Reconstitute antibody with 50 µL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Peptide sequence is: (C)EDKVRSL KTDIEESK
Homology: Mouse, human amino acid residues are identical.
This gene encodes a member of the homer family of dendritic proteins. Members of this family regulate group 1 metabotrophic glutamate receptor function. The encoded protein is a postsynaptic density scaffolding protein. Alternative splicing results in multiple transcript variants. Two related pseudogenes have been identified on chromosome 14.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A VESL-2; Cupidin; homer homolog 2; homer homolog 3; Homer protein homolog 2; homer, neuronal immediate early gene, 2; Homer-2; VASP/Ena-related gene up-regulated during seizure and LTP 2; Vesl-2
Gene Aliases: 9330120H11Rik; ACPD; AW539445; CPD; DFNA68; HOMER-2; HOMER2; VESL-2; Vesl2
UniProt ID: (Human) Q9NSB8, (Mouse) Q9QWW1, (Rat) O88801
Entrez Gene ID: (Human) 9455, (Mouse) 26557, (Rat) 29547
Molecular Function:
scaffold/adaptor protein
immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support