Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MATAQLQRTP MSALVFPNKI STEHQSLVLV KRLLAVSVSC ITYLRGIFPE CAYGTRYLDD LCVKILREDK NCPGSTQLVK WMLGCYDALQ KKYLRMVVLA VYTNPEDPQT ISECYQFKFK YTNNGPLMDF ISKNQSNESS MLSTDTKKAS ILLIRKIYIL MQNLGPLPND VCLTMKLFYY DEVTPPDYQP PGFKDGDCEG VIFEGEPMYL NVGEVSTPFH IFKVKVTTER ERMENIDSTI LSPKQIKTPF QKILRDKDVE DEQEHYTSDD LDIETKMEEQ EKNPASSELE EPSLVCEEDE IMRSKESPDL SISHSQVEQL VNKTSELDMS ESKTRSGKVF QNKMANGNQP VKSSKENRKR SQHESGRIVL HHFDSSSQES VPKRRKFSEP KEHI
This gene encodes a HORMA domain-containing protein. HORMA domains are involved in chromatin binding and play a role in cell cycle regulation. The encoded protein may play a role in meiosis, and expression of this gene is a potential marker for cancer. A pseudogene of this gene is located on the long arm of chromosome 6. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 46; DKFZP434A1315; HORM1; HORMA domain containing protein; newborn ovary HORMA protein; testis tissue sperm-binding protein Li 86P; unnamed protein product
Gene Aliases: CT46; NOHMA
UniProt ID: (Human) Q86X24
Entrez Gene ID: (Human) 84072
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support