Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Biologo
Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Paraffin) (IHC (P)) |
Assay-dependent | - |
Immunohistochemistry (Frozen) (IHC (F)) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Virus |
Host/Isotype |
Mouse / IgG |
Class |
Monoclonal |
Type |
Antibody |
Clone |
1H8 |
Immunogen |
SDS-disrupted bovine papillomavirus type 1 |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
30 µL/Test |
Purification |
Protein A/G |
Storage buffer |
PBS |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
Positive control: Infected cervical biopsy.
Human Papilloma virus (HPV) is responsible for the development of type and tissue specific papilloma in the oral cavity and in the larynx. In the skin papilloma virus can cause different types of warts and in the ano-genital region it is associated with condyloma or carcinoma. HPV types 1, 6 and 11 usually are associated with benign transformations of the tissues, while e.g. HPV types 16, 18 and 31 seem to be responsible for the development of some carcinoma like cervical carcinoma. The role of HPV in non-melanoic tumours of the skin is under discussion. Papillomavirus Type 1, 6, 11, 16, 18, 31.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support