Search Thermo Fisher Scientific
Reactive with the L1 of types 6, 11, 16, 18, 31, 33, 45, 52, & 58
The papillomavirus major capsid protein, L1, is a ~55 kD protein with the ability to spontaneously self-assemble into virus-like particles (VLPs). These VLPs present an exterior surface essentially indistinguishable from the native 60 nm non-enveloped papillomavirus virion. Purified recombinant L1 proteins can achieve this complex assembly reaction in the absence of any chaperones (Schiller and Lowy, 2012). PMID: 23800545.
Protein Aliases: Human Papillomavirus; Human Papillomavirus L1; Human Papillomavirus L1 Pan