Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200-1:2,000 | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:150 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Product Specifications | |
---|---|
Species Reactivity |
Dog, Human, Mouse, Rat |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI1D9 |
Immunogen |
Full length human recombinant protein of human HRAS produced in HEK293T cell. |
Conjugate |
Unconjugated |
Form |
lyophilized |
Concentration |
1 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.3, with 8% trehalose |
Contains |
no preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Monomeric G protein Ras functions as a molecular switch linking receptor and nonreceptor tyrosine kinase activation to a number of downstream signaling pathways leading to distinct cytoplasmic or nuclear events. Each mammalian cell contains three Ras proto-oncogene coding for closely related Ras proteins: H-, K-, N-Ras. Oncogenic mutations in Ras genes are present in ~30% of all human cancers. Constitutive activation of Ras due to mutations or overexpression stimulates proliferation and inhibition of apoptosis. K-Ras mutations are common in pancreatic, colorectal and nonsmall-cell lung carcinomas; H-Ras mutations are common in bladder, kidney and thyroid carcinomas; N-Ras mutations are found in melanoma, hematological malignancies and hepatocellular carcinaoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-H-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; c-has/bas p21 protein; c-ras-Ki-2 activated oncogene; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase Hras1; H-ras 1 protein; H-Ras-1; Ha-Ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene 1; p19 H-RasIDX protein; p21ras; Ras family small GTP binding protein H-Ras; ras p21; transformation gene: oncogene HAMSV; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog
Gene Aliases: C-BAS/HAS; C-H-RAS; c-Ha-ras; C-HA-RAS1; c-rasHa; CTLO; H-RAS; H-RASIDX; Ha-ras; HAMSV; Harvey-ras; HRAS; Hras-1; HRAS1; Kras2; p21ras; ras; RASH1
UniProt ID: (Human) P01112, (Mouse) Q61411, (Rat) P20171
Entrez Gene ID: (Dog) 403735, (Human) 3265, (Mouse) 15461, (Rat) 293621
Molecular Function:
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support