Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Recommended positive controls: 293T, A431, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat-2, RK3E, 4T1, SW480.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (94%), Xenopus laevis (94%), Pig (100%), Chicken (94%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock 84 kDa; heat shock 90kD protein 1, beta; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, beta; heat shock protein 90 kDa; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; heat shock protein, 84 kDa 1; HSP; HSP 84; HSP 90; HSP84; HSP90-beta; retinal degeneration slow protein; RP1-302G2.1; TSTA; Tumor-specific transplantation 84 kDa antigen
Gene Aliases: 90kDa; AL022974; C81438; D6S182; HSP84; Hsp84-1; Hsp90; HSP90-BETA; HSP90AB1; HSP90B; HSPC2; HSPCB
UniProt ID: (Human) P08238, (Mouse) P11499, (Rat) P34058
Entrez Gene ID: (Human) 3326, (Mouse) 15516, (Rat) 301252
Molecular Function: Hsp90 family chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support