Invitrogen
HYB 267-01-02 has specificity for bovine serum albumin (BSA). This antibody reacts with human serum albumin (25%), but does not cross-react with rabbit serum albumin.
HYB 267-01-02 has been successfully used in ELISA and Western blotting techniques. HYB 267-01-02 reacts strongly with BSA. Strong reaction is seen in ELISA with bovine serum albumin coated directly onto the microtiter well. In Western blotting after SDS-PAGE, HYB 267-01-02 reacts with bovine albumin.
The HYB 267-01-02 immunogen is BSA isolated from bovine plasma.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
Protein Aliases: Albumin; albumin (32 AA); albumin (AA 34); Bos d 6; bovine albumin; BSA; cell growth inhibiting protein 42; growth-inhibiting protein 20; MGC179277; serum albumin
Gene Aliases: ALB; GIG20; GIG42; PRO0903; PRO1708; PRO2044; PRO2619; PRO2675; UNQ696/PRO1341