Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MAVTEPKWVS VWSRFLWVTL LSMVLGSLLA LLLPLGAVEE QCLAVLKGLY LLRSKPDRAQ HAATKCTSPS TELSITSRGA TLLVAKTKAS PAGKLEARAA LNQALEMKRQ GKREKAQKLF MHALKMDPDF VDALTEFGIF SEEDKDIIQA DYLYTRALTI SPYHEKALVN RDRTLPLVEE IDQRYFSIID SKVKKVMSIP KGNSALRRVM EETYYHHIYH TVAIEGNTLT LSEIRHILET RYAVPGKSLE EQNEVIGMHA AMKYINTTLV SRIGSVTISD VLEIHRRVLG YVDPVEAGRF RTTQVLVGHH IPPHPQDVEK QMQEFVQWLN SEEAMNLHPV EFAALAHYKL VYIHPFIDGN GRTSRLLMNL ILMQAGYPPI TIRKEQRSDY YHVLEAANEG DVRPFIRFIA KCTETTLDTL LFATTEYSVA LPEAQPNHSG FKETLPVKP
Protein that can both mediate the addition of adenosine 5'-monophosphate (AMP) to specific residues of target proteins (AMPylation), and the removal of the same modification from target proteins (de-AMPylation), depending on the context. The side chain of Glu-231 determines which of the two opposing activities (AMPylase or de-AMPylase) will take place. Acts as a key regulator of the ERN1/IRE1-mediated unfolded protein response (UPR) by mediating AMPylation or de-AMPylation of HSPA5/BiP (PubMed:25601083). In unstressed cells, acts as an adenylyltransferase by mediating AMPylation of HSPA5/BiP at 'Thr-518', thereby inactivating it. In response to endoplasmic reticulum stress, acts as a phosphodiesterase by mediating removal of ATP (de-AMPylation) from HSPA5/BiP at 'Thr-518', leading to restore HSPA5/BiP activity. Although it is able to AMPylate RhoA, Rac and Cdc42 Rho GTPases in vitro, Rho GTPases do not constitute physiological substrates (PubMed:19362538, PubMed:25601083). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenosine monophosphate-protein transferase FICD; AMPylase; AMPylator FICD; De-AMPylase FICD; FIC domain containing; FIC domain-containing protein; fic S-phase protein cell division homolog; HIP-13; Huntingtin interacting protein E; huntingtin interactor protein E; Huntingtin yeast partner E; Huntingtin-interacting protein 13; Huntingtin-interacting protein E; MGC5623; Protein adenylyltransferase FICD
Gene Aliases: FICD; HIP13; HYPE; SPG92; UNQ3041; UNQ3041/PRO9857
UniProt ID: (Human) Q9BVA6
Entrez Gene ID: (Human) 11153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support